Gene Transcripts

Gene Symbol
Catsper4
Entrez Gene ID
329954
Gene Name
cation channel, sperm associated 4
Gene Aliases
4933412L05
Chromosome Location
Chr. 4: 134211968 - 134227434 on Build GRCm38
UniGene ID
Mm.79072
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001130030.1 NP_001123502.1 3 320
NM_177866.4 NP_808534.1 3 320
GenBank mRNA AK077145.1 - 3 307
BC051630.1 - 3 308
BC108974.2 - 3 300 40047424
BC108975.1 - 3 300 40047425
BY104668.1 - 3 321
EU781510.1 - 3 295
EU781511.1 - 3 295

View Full Product Details