Gene Transcripts

Gene Symbol
ST8SIA4
Entrez Gene ID
7903
Gene Name
ST8 alpha-N-acetyl-neuraminide alpha-2,8-sialyltransferase 4
Gene Aliases
PST, PST1, SIAT8D, ST8SIA-IV
Chromosome Location
Chr. 5: 100806935 - 100903285 on Build GRCh38
UniGene ID
Hs.308628
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005668.5 NP_005659.1 2 512
NM_175052.2 NP_778222.1 2 512
XM_005272078.2 XP_005272135.1 2 512
XM_011543630.1 XP_011541932.1 2 512
GenBank mRNA AK292872.1 - 2 464
BC027866.1 - 2 441
BC053657.1 - 2 493 5752337
BI858673.1 - 2 508
BM921020.1 - 2 512
DA862968.1 - 2 512
L41680.1 - 2 394

View Full Product Details