Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001289458.1 | NP_001276387.1 | 4 | 547 | |
NM_001289459.1 | NP_001276388.1 | 4 | 536 | ||
NM_001289460.1 | NP_001276389.1 | 4 | 547 | ||
NM_001289461.1 | NP_001276390.1 | 3 | 369 | ||
NM_010427.5 | NP_034557.3 | 4 | 519 | ||
GenBank mRNA | AF042856.1 | - | 3 | 348 | |
AK042121.1 | - | 4 | 465 | ||
AK052289.1 | - | 4 | 548 | ||
AK082461.1 | - | 3 | 519 | ||
AK142159.1 | - | 4 | 465 | ||
BC119228.1 | - | 4 | 406 | 8733977 | |
D10212.1 | - | 3 | 342 | ||
D10213.1 | - | 3 | 342 | ||
S71816.1 | - | 3 | 342 | ||
X72307.1 | - | 3 | 403 | ||
X84046.1 | - | 3 | 382 |