Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001289458.1 | NP_001276387.1 | 6 | 791 | |
NM_001289459.1 | NP_001276388.1 | 6 | 780 | ||
NM_001289460.1 | NP_001276389.1 | 6 | 791 | ||
NM_001289461.1 | NP_001276390.1 | 5 | 598 | ||
NM_010427.5 | NP_034557.3 | 6 | 763 | ||
GenBank mRNA | AF042856.1 | - | 5 | 592 | |
AK042121.1 | - | 6 | 709 | ||
AK052289.1 | - | 6 | 792 | ||
BC119228.1 | - | 6 | 650 | 8733977 | |
BC144764.1 | - | 6 | 662 | ||
D10212.1 | - | 5 | 586 | ||
D10213.1 | - | 5 | 571 | ||
S71816.1 | - | 5 | 586 | ||
X72307.1 | - | 5 | 647 | ||
X84046.1 | - | 5 | 626 |