Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_026598.3 | NP_080874.2 | 4 | 657 | |
GenBank mRNA | AF243434.2 | - | 4 | 613 | |
AK011237.1 | - | 4 | 616 | ||
AK019140.1 | - | 4 | 509 | ||
AK019963.1 | - | 4 | 652 | ||
AK044618.1 | - | 2 | 1145 | ||
AK163371.1 | - | 4 | 616 | ||
BC027422.1 | - | 4 | 635 | 4035224 |