Gene Transcripts

Gene Symbol
Cdkal1
Entrez Gene ID
68916
Gene Name
CDK5 regulatory subunit associated protein 1-like 1
Gene Aliases
1190005B03Rik, 6620401C13Rik
Chromosome Location
Chr. 13: 29191751 - 29855684 on Build GRCm38
UniGene ID
Mm.212227
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK004490.1 - 1 31
AK018346.1 - 12 1202
AK082629.1 - 12 1204
AK158546.1 - 12 1194
BC016073.1 - 12 1145 4482130

View Full Product Details