Gene Transcripts

Gene Symbol
Olfr63
Entrez Gene ID
258939
Gene Name
olfactory receptor 63
Gene Aliases
M4, MOR267-1
Chromosome Location
Chr. 17: 33268726 - 33269676 on Build GRCm38
UniGene ID
Mm.377759
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146937.1 NP_667148.1 1 274
GenBank mRNA BC127997.1 - 1 278

View Full Product Details