Gene Transcripts

Gene Symbol
ARPC2
Entrez Gene ID
10109
Gene Name
actin related protein 2/3 complex subunit 2
Gene Aliases
ARC34, PNAS-139, PRO2446, p34-Arc
Chromosome Location
Chr. 2: 218217094 - 218254348 on Build GRCh38
UniGene ID
Hs.529303
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005731.3 NP_005722.1 4 315
NM_152862.2 NP_690601.1 5 410
XM_017003113.1 XP_016858602.1 4 284
XM_017003114.1 XP_016858603.1 5 357
GenBank mRNA AF006085.1 - 4 325
AF279893.1 - 3 167
AK092520.1 - 1 1372
AK290268.1 - 5 309
BC000590.2 - 5 273 3346685
BG282181.1 - 5 409
BG765898.1 - 5 353
BT006898.1 - 4 241
U50523.1 - 5 325

View Full Product Details