Gene Transcripts

Gene Symbol
Tmem82
Entrez Gene ID
213989
Gene Name
transmembrane protein 82
Gene Aliases
Chromosome Location
Chr. 4: 141614229 - 141618633 on Build GRCm38
UniGene ID
Mm.34891
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145987.2 NP_666099.2 4 1374
GenBank mRNA AK085940.1 - 1375
AK154809.1 - 3 1433
AK162322.1 - 4 726
BC025833.1 - 4 775 5125918

View Full Product Details