Gene Transcripts

Gene Symbol
Agbl1
Entrez Gene ID
244071
Gene Name
ATP/GTP binding protein-like 1
Gene Aliases
Ccp4, D430020F16, EG244071, Nna1-l1
Chromosome Location
Chr. 7: 76229887 - 77124698 on Build GRCm38
UniGene ID
Mm.322829
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001199224.1 NP_001186153.1 11 1532
GenBank mRNA AK052433.1 - 5 1018
DQ867033.1 - 5 1018
FN429927.1 - 11 1532

View Full Product Details