Gene Transcripts

Gene Symbol
Galntl6
Entrez Gene ID
270049
Gene Name
UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase-like 6
Gene Aliases
1700021K10Rik, 4930431L04Rik, A830023L05
Chromosome Location
Chr. 8: 57772489 - 58911627 on Build GRCm38
UniGene ID
Mm.236368
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175032.3 NP_778197.2 3 499
GenBank mRNA AK080614.1 - 3 499
BB641428.1 - 3 499
BC111102.1 - 4 542
GU220061.1 - 3 346

View Full Product Details