Gene Transcripts

Gene Symbol
ATP6AP2
Entrez Gene ID
10159
Gene Name
ATPase H+ transporting accessory protein 2
Gene Aliases
APT6M8-9, ATP6IP2, ATP6M8-9, ELDF10, HT028, M8-9, MRXE, MRXSH, MSTP009, PRR, RENR, XMRE, XPDS
Chromosome Location
Chr. X: 40580964 - 40606637 on Build GRCh38
UniGene ID
Hs.495960
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005765.2 NP_005756.2 2 186
GenBank mRNA AF109363.1 - 2 147
AF248966.1 - 2 191
AF291814.1 - 2 173
AK075382.1 - 2 244
AK293510.1 - 2 168
AK297007.1 - 2 157
AK297760.1 - 2 256
AK314633.1 - 2 137
AK315948.1 - 2 117
AL049929.1 - 1 26
AY038990.1 - 2 189
AY429341.1 - 1 49
BC084541.1 - 2 166 6575732

View Full Product Details