Gene Transcripts

Gene Symbol
Nek5
Entrez Gene ID
330721
Gene Name
NIMA (never in mitosis gene a)-related expressed kinase 5
Gene Aliases
6530411J22, BB049667
Chromosome Location
Chr. 8: 22073615 - 22125415 on Build GRCm38
UniGene ID
Mm.113940
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177898.4 NP_808566.2 3, 4 288
GenBank mRNA AK032672.1 - 3, 4 290
AK054168.1 - 3, 4 302
BC053516.1 - 3, 4 276 6311513

View Full Product Details