Gene Transcripts

Gene Symbol
Gprin3
Entrez Gene ID
243385
Gene Name
GPRIN family member 3
Gene Aliases
C030038J10Rik, C730021L23
Chromosome Location
Chr. 6: 59344503 - 59426290 on Build GRCm38
UniGene ID
Mm.138080
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_183183.2 NP_899006.1 2 1897
GenBank mRNA AK050148.1 - 2 1898
AK173336.1 - 2 1901
BC138149.1 - 1 1767 8861168
BC138150.1 - 1 1767 8861169

View Full Product Details