Gene Transcripts

Gene Symbol
Slfn5
Entrez Gene ID
327978
Gene Name
schlafen 5
Gene Aliases
AI956840
Chromosome Location
Chr. 11: 82951349 - 82964850 on Build GRCm38
UniGene ID
Mm.42147
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_183201.4 NP_899024.3 6 2839
GenBank mRNA AK036486.1 - 6 2840
BC117909.1 - 5 2719 40097236
BC117910.1 - 5 2719 40097237

View Full Product Details