Gene Transcripts

Gene Symbol
Zmiz1
Entrez Gene ID
328365
Gene Name
zinc finger, MIZ-type containing 1
Gene Aliases
BC065120, E330020C23, ENSMUSG00000072684, Gm10397, I920194n01, Rai17, Zimp10
Chromosome Location
Chr. 14: 25458865 - 25666747 on Build GRCm38
UniGene ID
Mm.227484
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_183208.3 NP_899031.2 7 1012
GenBank mRNA AK054366.1 - 7 906
BC058646.1 - 7 930 6405777
BC065120.1 - 7 905 6856060

View Full Product Details