Gene Transcripts

Gene Symbol
Tsen2
Entrez Gene ID
381802
Gene Name
tRNA splicing endonuclease 2 homolog (S. cerevisiae)
Gene Aliases
AU067695
Chromosome Location
Chr. 6: 115544704 - 115578340 on Build GRCm38
UniGene ID
Mm.291208
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_199033.1 NP_950198.1 2 158
GenBank mRNA AK044190.1 - 2 156
AK134697.1 - 2 154
BC061473.1 - 2 158 6414790

View Full Product Details