Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_199035.2 | NP_950200.2 | 11, 12 | 1302 | |
GenBank mRNA | AK141341.1 | - | 11, 12 | 1299 | |
AK166755.1 | - | 11, 12 | 1297 | ||
AK166897.1 | - | 11, 12 | 1303 | ||
BC061244.1 | - | 11, 12 | 1291 | 30246181 |