Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_021436.2 | NP_067411.1 | 3, 4 | 685 | |
GenBank mRNA | AJ400622.1 | - | 3, 4 | 351 | |
AK043792.1 | - | 1, 2 | 58 | ||
AK079633.1 | - | 1, 2 | 38 | ||
BC057598.1 | - | 3, 4 | 685 | 6401547 |