Gene Transcripts

Gene Symbol
Slc44a3
Entrez Gene ID
213603
Gene Name
solute carrier family 44, member 3
Gene Aliases
BC010552
Chromosome Location
Chr. 3: 121459528 - 121532344 on Build GRCm38
UniGene ID
Mm.33062
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145394.3 NP_663369.2 9 1085
GenBank mRNA BC010552.1 - 8 853
BC025548.1 - 9 1083 5320841

View Full Product Details