Gene Transcripts

Gene Symbol
Pdlim2
Entrez Gene ID
213019
Gene Name
PDZ and LIM domain 2
Gene Aliases
4732462F18Rik, Slim
Chromosome Location
Chr. 14: 70164218 - 70177703 on Build GRCm38
UniGene ID
Mm.283968
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001253736.1 NP_001240665.1 10 1365
NM_145978.2 NP_666090.1 10 1292
GenBank mRNA BC024556.1 - 10 1292

View Full Product Details