Gene Transcripts
- Gene Symbol
- Ddx27
- Entrez Gene ID
- 228889
- Gene Name
- DEAD (Asp-Glu-Ala-Asp) box polypeptide 27
- Gene Aliases
- C86129
- Chromosome Location
- Chr. 2: 167015255 - 167034950 on Build GRCm38
- UniGene ID
- Mm.295031
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC024730.1 |
- |
8 |
1546 |
|
BC024981.1 |
- |
8 |
1546 |
|
BC033398.1 |
- |
8 |
1546 |
|
View Full Product Details