Gene Transcripts

Gene Symbol
Fnbp1l
Entrez Gene ID
214459
Gene Name
formin binding protein 1-like
Gene Aliases
2610318I01Rik, AW548221, TOCA1, toca-1
Chromosome Location
Chr. 3: 122538719 - 122619714 on Build GRCm38
UniGene ID
Mm.209491
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001114665.2 NP_001108137.1 16 2653
NM_153118.3 NP_694758.2 14 2479
GenBank mRNA AK135825.1 - 1 34
BC034510.1 - 9 1927 4481144

View Full Product Details