Gene Transcripts

Gene Symbol
Wdr19
Entrez Gene ID
213081
Gene Name
WD repeat domain 19
Gene Aliases
C330027H04Rik, D330023L08Rik, DYF2, Ift144, PWDMP, mKIAA1638
Chromosome Location
Chr. 5: 65199658 - 65260415 on Build GRCm38
UniGene ID
Mm.127550
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153391.2 NP_700440.2 7 607
GenBank mRNA AK043836.1 - 7 564
AK052303.1 - 7 587
AK147970.1 - 7 607
AK166172.1 - 7 573
AY029258.1 - 7 677
BC060134.1 - 7 587
BC082301.1 - 6 487

View Full Product Details