Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_153541.3 | NP_705769.2 | 2 | 631 | |
GenBank mRNA | AK132152.1 | - | 2 | 669 | |
AK140914.1 | - | 2 | 620 | ||
AK163930.1 | - | 2 | 632 | ||
AK163989.1 | - | 2 | 635 | ||
BC023839.1 | - | 2 | 626 | 5344921 |