Gene Transcripts

Gene Symbol
Cxcl17
Entrez Gene ID
232983
Gene Name
chemokine (C-X-C motif) ligand 17
Gene Aliases
VCC-1, Vcc1
Chromosome Location
Chr. 7: 25400053 - 25412886 on Build GRCm38
UniGene ID
Mm.10545
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153576.2 NP_705804.2 4 455
GenBank mRNA AK143005.1 - 4 371
AY374329.1 - 4 358
AY598463.1 - 4 456
BC024561.1 - 4 456 5004999

View Full Product Details