Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001167976.1 | NP_001161448.1 | 5 | 317 | |
NM_172577.3 | NP_766165.2 | 4 | 760 | ||
GenBank mRNA | AK036988.1 | - | 4 | 500 | |
AK041694.1 | - | 4 | 761 | ||
AK044945.1 | - | 4 | 263 | ||
AK045180.1 | - | 4 | 586 | ||
AK162379.1 | - | 4 | 587 | ||
AK167535.1 | - | 4 | 468 | ||
BC057980.1 | - | 4 | 584 | 5042653 | |
EU180236.1 | - | 5 | 317 |