Gene Transcripts
- Gene Symbol
- Kctd3
- Entrez Gene ID
- 226823
- Gene Name
- potassium channel tetramerisation domain containing 3
- Gene Aliases
- 4930438A20Rik, 9330185B06, E330032J19Rik, NY-REN-45
- Chromosome Location
- Chr. 1: 188971095 - 189007868 on Build GRCm38
- UniGene ID
- Mm.209880
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_172650.2 |
NP_766238.1
|
6, 7 |
633 |
|
GenBank mRNA |
AK033491.1 |
- |
6, 7 |
634 |
|
AK034380.1 |
- |
6, 7 |
633 |
|
AK049362.1 |
- |
6, 7 |
631 |
|
AK087407.1 |
- |
1, 2 |
745 |
|
AK087865.1 |
- |
6, 7 |
634 |
|
AK139700.1 |
- |
6, 7 |
633 |
|
AK215667.1 |
- |
1, 2 |
46 |
|
BC061458.1 |
- |
6, 7 |
621 |
|
BC079581.1 |
- |
6, 7 |
620 |
5702630 |
View Full Product Details