Gene Transcripts

Gene Symbol
Lin54
Entrez Gene ID
231506
Gene Name
lin-54 homolog (C. elegans)
Gene Aliases
4832414L08, AI461788, mKIAA2037
Chromosome Location
Chr. 5: 100442033 - 100500649 on Build GRCm38
UniGene ID
Mm.212568
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001115010.1 NP_001108482.1 12, 13 2355
NM_172714.4 NP_766302.2 12, 13 2223
GenBank mRNA AK029288.1 - 13, 14 1669
AK030894.1 - 12, 13 2357
AK134320.1 - 12, 13 2221
AK220225.1 - 12, 13 2218
BC125580.1 - 10, 11 1350
BC145025.1 - 10, 11 1350 9053573

View Full Product Details