Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_173765.3 | NP_776126.1 | 3 | 429 | |
GenBank mRNA | AB095954.1 | - | 3 | 446 | |
AK038779.1 | - | 4 | 439 | ||
AK043807.1 | - | 3 | 407 | ||
AK136849.1 | - | 3 | 427 | ||
AK148405.1 | - | 3 | 428 | ||
BC094507.1 | - | 2 | 206 | ||
BC128330.1 | - | 2 | 301 | 40110405 | |
BY741513.1 | - | 3 | 430 |