Gene Transcripts

Gene Symbol
KLHL41
Entrez Gene ID
10324
Gene Name
kelch like family member 41
Gene Aliases
KBTBD10, Krp1, SARCOSIN
Chromosome Location
Chr. 2: 169509702 - 169526262 on Build GRCh38
UniGene ID
Hs.50550
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006063.2 NP_006054.2 1 203
GenBank mRNA AF056929.1 - 1 173
AF333387.1 - 1 203
AK316571.1 - 1 200
BC006534.2 - 1 181 2959774

View Full Product Details