Gene Transcripts
- Gene Symbol
- Dcun1d1
- Entrez Gene ID
- 114893
- Gene Name
- DCN1, defective in cullin neddylation 1, domain containing 1 (S. cerevisiae)
- Gene Aliases
- Rp42, SCCRO, Tes3, pTes3
- Chromosome Location
- Chr. 3: 35892103 - 35937605 on Build GRCm38
- UniGene ID
- Mm.374810
- Species
- Mus musculus
- Gene Symbol
- Tes3-ps
- Entrez Gene ID
- 54339
- Gene Name
- testis derived transcript 3, pseudogene
- Gene Aliases
- Chromosome Location
- Chr. 13: 49493652 - 49496389 on Build GRCm38
- UniGene ID
- Mm.441473
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK029314.1 |
- |
1 |
538 |
|
View Full Product Details