Gene Transcripts

Gene Symbol
Fgfrl1
Entrez Gene ID
116701
Gene Name
fibroblast growth factor receptor-like 1
Gene Aliases
FGFR5, FGFR5beta, FGFR5gamma
Chromosome Location
Chr. 5: 108694229 - 108706950 on Build GRCm38
UniGene ID
Mm.35691
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001164259.1 NP_001157731.1 4 882
NM_054071.2 NP_473412.1 5 1155
GenBank mRNA AF321300.1 - 5 882
AF321301.1 - 4 843
AJ293947.1 - 4 788
BC058745.1 - 4 757 6439421

View Full Product Details