Gene Transcripts

Gene Symbol
Otud7a
Entrez Gene ID
170711
Gene Name
OTU domain containing 7A
Gene Aliases
AJ430384, Otud7
Chromosome Location
Chr. 7: 63444745 - 63759905 on Build GRCm38
UniGene ID
Mm.156872
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_130880.1 NP_570950.1 4 476
GenBank mRNA AJ430384.1 - 4 476

View Full Product Details