Gene Transcripts

Gene Symbol
Bbc3
Entrez Gene ID
170770
Gene Name
BCL2 binding component 3
Gene Aliases
PUMA, PUMA/JFY1
Chromosome Location
Chr. 7: 16308321 - 16318334 on Build GRCm38
UniGene ID
Mm.7660
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133234.2 NP_573497.1 4 1576
GenBank mRNA AF332560.1 - 4 1520
BC044782.2 - 4 1541
BC060370.1 - 4 1506

View Full Product Details