Gene Transcripts

Gene Symbol
Fam19a5
Entrez Gene ID
106014
Gene Name
family with sequence similarity 19, member A5
Gene Aliases
TAFA5, Tafa-5, Tara-5
Chromosome Location
Chr. 15: 87544299 - 87759364 on Build GRCm38
UniGene ID
Mm.244319
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001252310.1 NP_001239239.1 2 332
NM_134096.2 NP_598857.1 2 284
GenBank mRNA AK090194.1 - 2 158
AK090307.1 - 2 334
AY325124.1 - 2 285
BC015306.1 - 2 285 4223181
BU057253.1 - 2 288
BY733542.1 - 2 334

View Full Product Details