Gene Transcripts

Gene Symbol
Pkhd1l1
Entrez Gene ID
192190
Gene Name
polycystic kidney and hepatic disease 1-like 1
Gene Aliases
AB055648, PKHDL1
Chromosome Location
Chr. 15: 44457371 - 44601376 on Build GRCm38
UniGene ID
Mm.211494
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138674.2 NP_619615.2 2 140
GenBank mRNA AB055648.1 - 2 140
AY219182.1 - 2 140

View Full Product Details