Gene Transcripts

Gene Symbol
Sectm1a
Entrez Gene ID
209588
Gene Name
secreted and transmembrane 1A
Gene Aliases
BC010462
Chromosome Location
Chr. 11: 121067395 - 121097213 on Build GRCm38
UniGene ID
Mm.69553
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145373.2 NP_663348.2 5 597
GenBank mRNA AK082020.1 - 4 289
BC010462.1 - 5 564 3585040

View Full Product Details