Gene Transcripts

Gene Symbol
Sectm1a
Entrez Gene ID
209588
Gene Name
secreted and transmembrane 1A
Gene Aliases
BC010462
Chromosome Location
Chr. 11: 121067395 - 121097213 on Build GRCm38
UniGene ID
Mm.69553
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145373.2 NP_663348.2 6 826
GenBank mRNA AK082020.1 - 5 518
AK170159.1 - 5 404
AK171246.1 - 5 410
BC010462.1 - 6 793 3585040

View Full Product Details