Gene Transcripts

Gene Symbol
Ido2
Entrez Gene ID
209176
Gene Name
indoleamine 2,3-dioxygenase 2
Gene Aliases
AI265623, C230043N17Rik, Ido-2, Indol1
Chromosome Location
Chr. 8: 24531892 - 24576404 on Build GRCm38
UniGene ID
Mm.219580
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145949.2 NP_666061.3 8 771
GenBank mRNA AK082385.1 - 7 962
BC026393.1 - 8 771 4195134
EF137182.1 - 8 771
EU440733.1 - 7 563

View Full Product Details