Gene Transcripts

Gene Symbol
Ints9
Entrez Gene ID
210925
Gene Name
integrator complex subunit 9
Gene Aliases
BC028953, D14Ertd231e
Chromosome Location
Chr. 14: 64950045 - 65039835 on Build GRCm38
UniGene ID
Mm.71332
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001253731.1 NP_001240660.1 2 376
NM_153414.4 NP_700463.2 2 344
GenBank mRNA AK035769.1 - 2 88
AK038979.1 - 2 77
AK077634.1 - 2 345
AK136992.1 - 2 71
AK150195.1 - 2 71
BC028953.1 - 1 34 5362016
BC055700.1 - 2 43 6808816
BY195822.1 - 2 109

View Full Product Details