Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001166619.1 | NP_001160091.1 | 1 | 188 | |
NM_001166620.1 | NP_001160092.1 | 1 | 188 | ||
NM_174993.2 | NP_778158.2 | 1 | 188 | ||
GenBank mRNA | AK076190.1 | - | 1 | 146 | |
AY214130.1 | - | 1 | 93 | ||
BC099411.1 | - | 1 | 188 | 30788714 | |
BC138087.1 | - | 1 | 169 | 8861106 | |
BC145139.1 | - | 1 | 169 | 9053689 | |
BC145140.1 | - | 1 | 169 | 9053690 | |
BC171922.1 | - | 1 | 169 | 9053688 | |
BC171923.1 | - | 1 | 169 | 9053691 | |
BC171947.1 | - | 1 | 169 | 9054233 | |
BC171948.1 | - | 1 | 169 | 9054234 |