Gene Transcripts
- Gene Symbol
- Klk9
- Entrez Gene ID
- 101533
- Gene Name
- kallikrein related-peptidase 9
- Gene Aliases
- 1200016C12Rik, AI324041
- Chromosome Location
- Chr. 7: 43791588 - 43796758 on Build GRCm38
- UniGene ID
- Mm.19942
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK004807.1 |
- |
5 |
706 |
|
View Full Product Details