Gene Transcripts
- Gene Symbol
- Trappc9
- Entrez Gene ID
- 76510
- Gene Name
- trafficking protein particle complex 9
- Gene Aliases
- 1810044A24Rik, 2900005P22Rik, 4632408O18Rik, Ibp, Nibp, TRS130, mKIAA1882
- Chromosome Location
- Chr. 15: 72589620 - 73061243 on Build GRCm38
- UniGene ID
- Mm.158902
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001164641.1 |
NP_001158113.1
|
9, 10 |
1755 |
|
NM_001164642.1 |
NP_001158114.1
|
8, 9 |
1728 |
|
NM_029640.2 |
NP_083916.1
|
8, 9 |
983 |
|
NM_180662.2 |
NP_850993.2
|
8, 9 |
1728 |
|
GenBank mRNA |
AK028504.1 |
- |
8, 9 |
1570 |
|
AK029106.1 |
- |
9, 10 |
1597 |
|
AK138682.1 |
- |
8, 9 |
1575 |
|
AK156739.1 |
- |
8, 9 |
1553 |
|
AY630620.1 |
- |
8, 9 |
981 |
|
BC034590.1 |
- |
8, 9 |
981 |
5364921 |
BC038938.1 |
- |
2, 3 |
298 |
|
BC070463.1 |
- |
8, 9 |
1728 |
30613261 |
View Full Product Details