Gene Transcripts

Gene Symbol
Lcn9
Entrez Gene ID
77704
Gene Name
lipocalin 9
Gene Aliases
9230102I19Rik
Chromosome Location
Chr. 2: 25823094 - 25825542 on Build GRCm38
UniGene ID
Mm.159974
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029959.2 NP_084235.1 4 356
GenBank mRNA AF435738.1 - 4 356
AK020305.1 - 4 358

View Full Product Details