Gene Transcripts

Gene Symbol
Pcdhb22
Entrez Gene ID
93893
Gene Name
protocadherin beta 22
Gene Aliases
Pcdhb15, PcdhbV
Chromosome Location
Chr. 18: 37518353 - 37521419 on Build GRCm38
UniGene ID
Mm.348054
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053147.3 NP_444377.3 1 2979
GenBank mRNA AK043492.1 - 1 2980

View Full Product Details