Gene Transcripts

Gene Symbol
Ankrd10
Entrez Gene ID
102334
Gene Name
ankyrin repeat domain 10
Gene Aliases
4833425P12Rik, AW549277
Chromosome Location
Chr. 8: 11611581 - 11636136 on Build GRCm38
UniGene ID
Mm.12459
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001167967.2 NP_001161439.1 2 389
NM_001281974.1 NP_001268903.1 2 389
NM_001281975.1 NP_001268904.1 2 389
NM_133971.3 NP_598732.2 2 389
GenBank mRNA AK014768.1 - 2 268
AK031808.1 - 2 387
AK033623.1 - 2 387
AK038156.1 - 2 387
AK042500.1 - 2 387
AK050983.1 - 2 390
AK081754.1 - 2 387
AK082875.1 - 2 386
AK084456.1 - 2 387
AK088371.1 - 2 339
AK143253.1 - 2 387
BC002198.1 - 2 321 3489133

View Full Product Details