Gene Transcripts

Gene Symbol
Sfxn5
Entrez Gene ID
94282
Gene Name
sideroflexin 5
Gene Aliases
C230001H08Rik
Chromosome Location
Chr. 6: 85213049 - 85333483 on Build GRCm38
UniGene ID
Mm.121485
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178639.4 NP_848754.1 14 1729
GenBank mRNA AK078142.1 - 1 627
AK082073.1 - 14 1729

View Full Product Details