Gene Transcripts

Gene Symbol
Trim68
Entrez Gene ID
101700
Gene Name
tripartite motif-containing 68
Gene Aliases
AI894158, F730114J12Rik, Rnf137, SS-56
Chromosome Location
Chr. 7: 102677579 - 102687329 on Build GRCm38
UniGene ID
Mm.39043
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK164544.1 - 4 847
BC034249.1 - 4 830 3675490
CO424835.1 - 2 309

View Full Product Details