Gene Transcripts

Gene Symbol
Rab35
Entrez Gene ID
77407
Gene Name
RAB35, member RAS oncogene family
Gene Aliases
9530019H02Rik, AU040256, H-ray, RAB1C, RAY
Chromosome Location
Chr. 5: 115631882 - 115647158 on Build GRCm38
UniGene ID
Mm.27553
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198163.1 NP_937806.1 2 236
GenBank mRNA AB232631.1 - 2 76
AK156576.1 - 2 227
BC056466.1 - 2 236 5717246

View Full Product Details